missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BES, 99+%, for biochemistry

Product Code. p-65622 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10082210

Brand: Thermo Scientific Chemicals 172580250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10191-18-1 | C6H15NO5S | 213.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10191-18-1
Molecular Formula C6H15NO5S
Molecular Weight (g/mol) 213.25
MDL Number MFCD00007533
InChI Key AJTVSSFTXWNIRG-UHFFFAOYSA-N
Synonym N, N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid
PubChem CID 73243
ChEBI CHEBI:39045
SMILES OCCN(CCO)CCS(O)(=O)=O

Specifications

Absorbance 0.095 max. (1 M in H2O) at 260nm
Melting Point 152.0°C to 154.0°C
CAS Min % 99.0
CAS Max % 100.0
Color Beige to Brown
Physical Form Powder
Quantity 25 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (100°C, 2 hrs)
Linear Formula (HOCH2CH2)2NCH2CH2SO3H
Packaging Glass bottle
Solubility Information Solubility in water: soluble.
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Formula Weight 213.25
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material BES
Show More Show Less
Product Identifier
  • BES

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.