missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-2 Microglobulin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA546943
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
beta-2 Microglobulin Polyclonal antibody specifically detects beta-2 Microglobulin in Rat samples. It is validated for Flow Cytometry, Western Blot.Specifications
beta-2 Microglobulin | |
Polyclonal | |
Unconjugated | |
B2m | |
B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m | |
Goat | |
Antigen affinity chromatography | |
RUO | |
24223 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P07151 | |
B2m | |
Mouse myeloma cell line NS0-derived recombinant rat beta 2-Microglobulin Ile21-Met119 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |