Learn More
Invitrogen™ beta-3 Defensin Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Brand: Invitrogen™ 900-M210
This item is not returnable.
View return policy
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
P81534 | |
414325 | |
2 x 96 Tests | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
DEFB103A | |
RUO | |
Human |