Learn More
beta-3 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£165.00 - £1374.00
Specifications
Antigen | beta-3 Defensin |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17865453
|
Invitrogen
500-P241-50UG |
50μg |
£165.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17855083
|
Invitrogen
500-P241-100UG |
100 μg |
£212.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17804913
|
Invitrogen
500-P241-1MG |
1 mg |
£1374.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-3. Anti-Human BD-3-specific antibody was purified by affinity chromatography employing an immobilized Human BD-3 matrix. Sandwich ELISA: To detect Human BD-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-3 (500-P241Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-3. Western Blot: To detect Human BD-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].Specifications
beta-3 Defensin | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P81534 | |
414325 | |
E.coli-derived Recombinant Human BD-3 | |
Antigen affinity chromatography | |
DEFB103A |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
DEFB103A | |
Primary | |
-20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.