Learn More
beta Actin Loading Control Monoclonal Antibody (BA3R), DyLight™ 800 4X PEG, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515739D800
Additional Details : Weight : 0.01000kg
Description
MA5-15739-D800 has been successfully used for Western blotting and immunofluorescence. DyLight 800 4X PEG has an excitation/emission of 783/797 nm. For visualization of actin filaments by immunofluorescence, fixation and permeabilization with methanol is required.
beta Actin Loading Control Monoclonal antibody specifically detects beta Actin Loading Control in Chicken, Human, Mouse, Rabbit, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
beta Actin Loading Control | |
Monoclonal | |
1 mg/mL | |
MES with proprietary stabilizer and no preservative | |
P29751, P60706, P60709, P60710, P60711 | |
ACTB | |
Beta-actin N-terminal peptide. | |
50 μL | |
Primary | |
Chicken, Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
BA3R | |
DyLight 800 4X PEG | |
ACTB | |
0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; acti | |
Mouse | |
Protein A | |
RUO | |
100009272, 11461, 396526, 60, 81822 | |
4° C, store in dark | |
Liquid |