Learn More
beta Actin Mouse anti-Hamster, Human, Mouse, Non-human primate, Rat, Zebrafish, Clone: 2D4H5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66009-1-IG-150UL
Product Code. 16817505
  • £189.00 / 150µL
Explore more special offers
Description
Description
This antibody can recognize all the actins. The isotype of this antibody is IgG2b.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Specifications
Specifications
beta Actin
Monoclonal
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48975, P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136
ACTA1, acta1b, acta2, ACTB, actb1, ACTC1, actc1b, Actg1, ACTG2
Protein A
RUO
100689477, 100758575, 100764022, 100766388, 100768744, 100768972, 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 407658, 57934, 58, 58114, 59, 60, 70, 71, 72, 81633, 81822
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
2D4H5
Unconjugated
ACTA1
ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin, PS1TP5BP1
Mouse
150 μL
Primary
Hamster, Human, Mouse, Non-human Primate, Rat, Zebrafish
Antibody
IgG2b