missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Allopyranose, 97%, Thermo Scientific™

Product Code. 15447429
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7283-09-2
MDL Number MFCD00063268

Specifications

Melting Point 148°C to 150°C
Color White
Molecular Formula C9H10ClNO2
Quantity 250 mg
Synonym 4-chlorophenyl dimethylcarbamate, p-chlorophenyl-n,n-dimethyl carbamate, 4-chlorophenyl n,n-dimethylcarbamate, dimethylcarbamic acid 4-chlorophenyl ester, n,n-dimethyl-4-chlorophenylcarbamate
InChI Key QTILATGINOFTLP-UHFFFAOYSA-N
SMILES CN(C)C(=O)OC1=CC=C(C=C1)Cl
IUPAC Name (4-chlorophenyl) N,N-dimethylcarbamate
Molecular Weight (g/mol) 199.63
PubChem CID 350036
Formula Weight 180.16
Percent Purity 97%
Physical Form Powder
Chemical Name or Material beta-D-Allopyranose
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'