missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals beta-D-Glucosamine pentaacetate, 96% Product Code.: 11382836

Thermo Scientific Chemicals beta-D-Glucosamine pentaacetate, 96%

Product Code. 11382836
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11382836

Brand: Thermo Scientific Chemicals L09020.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7772-79-4
Molecular Formula C16H23NO10
Molecular Weight (g/mol) 389.357
MDL Number MFCD00006595
InChI Key OVPIZHVSWNOZMN-OXGONZEZSA-N
Synonym n-acetyl-beta-d-glucosamine tetraacetate, 2-acetamido-1,3,4,6-tetra-o-acetyl-2-deoxy-beta-d-glucopyranose, beta-d-glucosamine pentaacetate, 2-acetamido-2-deoxy-beta-d-glucopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-5-acetamido-3,4,6-triacetoxy-tetrahydropyran-2-yl methyl acetate, beta-d-galactosamine pentaacetate, 2-acetylamino-2-deoxy-beta-d-galactopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-3,4,6-tris acetyloxy-5-acetamidooxan-2-yl methyl acetate, n-acetyl-, a-d-glucosamine tetraacetate, glc-n-ac free
PubChem CID 99461
IUPAC Name [(2R,3S,4R,5R,6S)-5-acetamido-3,4,6-triacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)NC1C(C(C(OC1OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 184°C to 189°C
Quantity 5 g
Beilstein 98835
Optical Rotation +4° (c=10 in Chloroform)
Formula Weight 389.37
Percent Purity 96%
Chemical Name or Material beta-D-Glucosamine pentaacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS06I022 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL09020.03,L09020.06
Lot NumberM30J005 Certificate TypeCertificate of Analysis Date03/02/2023 Catalog NumberL09020.03,L09020.06
Lot NumberU09C029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL09020.03,L09020.06
Lot NumberS06D038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL09020.03,L09020.06
Lot NumberT22A055 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL09020.03,L09020.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals beta-D-Glucosamine pentaacetate, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.