missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ beta-Galactosidase Assay Stop Solution

The Thermo Scientific Mammalian β-Galactosidase Assay Stop Solution is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

Brand:  Thermo Scientific™ 75706

Product Code. 11517830

  • £58.00 / 25mL
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Thermo Scientific Mammalian β-Galactosidase Assay Stop Solution is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Features of the Thermo Scientific Mammalian β-Galactosidase Assay Kit (assay reagent and stop solution):

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405 nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatiible with prepared lysates from other species such as bacteria

Related Products

  • Mammalian beta-Galactosidase Assay Kit (Cat. No. 75707)
  • beta-Galactosidase Assay Reagent (Cat. No. 75705)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-Galactosidase Assay Stop Solution, 25mL, 11.05pH, Liquid, Clear, 96-Well Plate, Reporter Enzyme Assay Type, Colorimetric Detection Method, Mammalian Compatible Cells, Proprietary Substrate, Beta-Gal Substrate, Absorbance Technique
Liquid
Reporter Enzyme
25mL
β-Galactosidase Assay Stop Solution, beta-Galactosidase Assay Stop Solution
96-well Plate
Pierce™
Beta-Gal Substrate
Store at room temperature.
Protein biology, enzyme reporter assays, protein assays and analysis, beta-galactosidase assays, Absorbance
11.05
Microplate Reader
Enzyme Labeled
25mL, 25 mL
Proprietary
Beta-Galactosidase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.