missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta Galactosidase Synthetic Peptide

Synthetic Peptide

Brand:  Invitrogen™ PEP0855

Product Code. 15359090

  • £127.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

beta-Galactosidase Synthetic Peptide, PEP-0855, from Invitrogen.

This peptide corresponds to 17 amino acids near the amino terminus of E. coli beta-galactosidase.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bacteria
beta Galactosidase
-20°C
Liquid
Blocking Assay, Control
0.02% sodium azide
50 μg
PBS
purified
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
No
Peptide
RUO - research use only
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers