Learn More
beta-Glycerophosphate, 200mM soln., Thermo Scientific Chemicals
£51.56 - £80.70
Chemical Identifiers
CAS | 819-83-0 |
---|---|
Molecular Formula | C3H9Na2O6P |
Molecular Weight (g/mol) | 218.052 |
MDL Number | MFCD00002135 |
InChI Key | UQVSFONYLAZJDB-UHFFFAOYSA-N |
Synonym | glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2oShow More |
PubChem CID | 131860121 |
IUPAC Name | 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium |
SMILES | C(C(CO)OP(=O)(O)O)O.[Na].[Na] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434559
|
Thermo Scientific Alfa Aesar
J62121.AD |
50 mL |
£51.56
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444559
|
Thermo Scientific Alfa Aesar
J62121.AE |
100 mL |
£80.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
β-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsβ-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
819-83-0 | |
218.052 | |
UQVSFONYLAZJDB-UHFFFAOYSA-N | |
131860121 | |
C(C(CO)OP(=O)(O)O)O.[Na].[Na] |
C3H9Na2O6P | |
MFCD00002135 | |
glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2oShow More | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium |
Specifications
819-83-0 | |
C3H9Na2O6P | |
glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o | |
UQVSFONYLAZJDB-UHFFFAOYSA-N | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium | |
131860121 | |
200mM soln. | |
beta-Glycerophosphate |
Colorless | |
MFCD00002135 | |
Soluble in water. | |
C(C(CO)OP(=O)(O)O)O.[Na].[Na] | |
218.052 | |
216.04 | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.