missing translation for 'onlineSavingsMsg'
Learn More
Learn More
β-Glycerophosphoric acid, disodium salt pentahydrate, 98%, Thermo Scientific Chemicals
beta-Glycerophosphoric acid, disodium salt pentahydrate, 98%, C3H7Na2O6P.5H2O, CAS Number-13408-09-8
£28.44 - £238.19
Chemical Identifiers
CAS | 13408-09-8 |
---|---|
Molecular Formula | C3H19Na2O11P |
Molecular Weight (g/mol) | 308.13 |
MDL Number | MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 |
InChI Key | VWVLGHQKEJESPW-UHFFFAOYSA-N |
Synonym | unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate |
PubChem CID | 131846916 |
IUPAC Name | 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate |
SMILES | O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10424701
|
Thermo Scientific Acros
410990250 |
25 g | Glass bottle |
£28.44
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259600
|
Thermo Scientific Acros
410991000 |
100 g | Glass bottle |
£68.44
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10561814
|
Thermo Scientific Acros
410995000 |
500 g | Glass bottle |
£238.19
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13408-09-8 | |
308.13 | |
VWVLGHQKEJESPW-UHFFFAOYSA-N | |
131846916 | |
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O |
C3H19Na2O11P | |
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate |
Specifications
13408-09-8 | |
100.0 | |
White | |
98% | |
CH2OHCH(OPO3Na2)CH2OH·5H2O | |
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate | |
VWVLGHQKEJESPW-UHFFFAOYSA-N | |
1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium;pentahydrate | |
131846916 | |
98% | |
β-Glycerophosphoric acid, disodium salt pentahydrate |
97.5 | |
102.0°C to 104.0°C | |
Authentic | |
C3H19Na2O11P | |
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135 | |
Solubility in water: freely soluble. | |
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O | |
308.13 | |
306.11 | |
Crystalline Powder |
Safety and Handling
RUO – Research Use Only