Learn More
Fisherbrand™ Beta Shields
Cleaver Scientific radiation shielding for protection from Beta radiation.
£304.00 - £416.00
Specifications
Product Type Beta Shield
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
12367177
View Documents
Fisherbrand™
CSR-S1T
15° Angle
£304.00
Each
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
12347177
View Documents
Fisherbrand™
CSR-S1OT
45° Angle
£416.00
Each
Estimated Shipment: 06-08-2025
to see stock.
Add to basket
Description
Description
  • Beta Shield - Small Fixed 15° Angle - Curved Base 30 x 45cm
  • Supplied in small, medium and large sizes and with a 15cm-deep curved base for use with safety trays, or a 30x30cm flat base for under-the-bench protection
  • A range of angles offers increased manoeuvrability, while clear optical acrylic aids visualisation
  • Radiation shielding Cleaver Scientific for protection from Beta radiation, shield, fixed 15 degree angle, curved base, dimensions 450h x 300w x 150d mm
Specifications
Specifications
Beta Shield
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Beta Shields