missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 18 Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
£330.00 - £423.30
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
11508730
|
Thermo Scientific™
71505-014006 |
4 mm | 10 mm | 5 μm |
£423.30
Pack of 4 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
11563251
|
Thermo Scientific™
71505-022106 |
2.1 mm | 20 mm | 5 μm |
£423.30
Pack of 4 |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11588720
|
Thermo Scientific™
71505-012106 |
2.1 mm | 10 mm | 5 μm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.
Specifications
5 μm | |
BetaBasic 18 | |
Beta |