missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals Betaine 5M (fr)
PCR grade betaine solution.
Brand: MP Biomedicals 11BETA0020
Additional Details : Weight : 0.00200kg
Description
- PCR grade.
- Half life: 1 year.
- Applications: PCR reagent.
- Non-sterile.
- Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Specifications
Store at +4°C | |
117.15 g/mol | |
(CH3)3N+CH2COO− | |
Solution |
Half life: 1 year | |
PCR reagent, Molecular biology | |
2 mL |