Learn More
Betaine, anhydrous, 98%, Thermo Scientific Chemicals
£31.40 - £342.00
Chemical Identifiers
CAS | 107-43-7 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.148 |
MDL Number | MFCD00012123 |
InChI Key | KWIUHFFTVRNATP-UHFFFAOYSA-N |
Synonym | betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin |
PubChem CID | 247 |
ChEBI | CHEBI:17750 |
IUPAC Name | 2-(trimethylazaniumyl)acetate |
SMILES | C[N+](C)(C)CC(=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405847
|
Thermo Scientific Alfa Aesar
B24397.22 |
100 g |
£31.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11415847
|
Thermo Scientific Alfa Aesar
B24397.36 |
500 g |
£88.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11425847
|
Thermo Scientific Alfa Aesar
B24397.0E |
2500 g |
£342.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Betaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBetaine is an active ingredient in toothpaste to control the symptoms of dryness of the mouth. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis. It is also used for boosting the immune system and for improving athletic performance. It is helpful to prevent noncancerous tumors in the colon (colorectal adenomas).
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
107-43-7 | |
117.148 | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
247 | |
2-(trimethylazaniumyl)acetate |
C5H11NO2 | |
MFCD00012123 | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
CHEBI:17750 | |
C[N+](C)(C)CC(=O)[O-] |
Specifications
107-43-7 | |
0.98 | |
MFCD00012123 | |
Hygroscopic | |
betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin | |
KWIUHFFTVRNATP-UHFFFAOYSA-N | |
2-(trimethylazaniumyl)acetate | |
247 | |
117.15 | |
Betaine, anhydrous |
>300°C (decomposition) | |
C5H11NO2 | |
3537113 | |
14,1179 | |
Soluble in water and methanol. | |
C[N+](C)(C)CC(=O)[O-] | |
117.148 | |
CHEBI:17750 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 203-490-6
missing translation for 'rtecsNumber' : DS5900000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only