missing translation for 'onlineSavingsMsg'
Learn More

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, Thermo Scientific Chemicals

Product Code. 10783541
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-46-5
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00011903
InChI Key HOPSCVCBEOCPJZ-UHFFFAOYSA-N
Synonym betaine hydrochloride, betaine chloride, pluchine, acidogeno, acipepsol, achylin, acidin, acinorm, acidol, acidine
PubChem CID 11545
IUPAC Name carboxymethyl(trimethyl)azanium;chloride
SMILES [Cl-].C[N+](C)(C)CC(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 241.0°C
Color Colorless to White
pH 0.8 to 1.2 (1:4)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3)3NCH2CO2H·Cl
Quantity 1 kg
Merck Index 15, 1181
Solubility Information Solubility in water: 647g/L (25°C). Other solubilities: soluble in ethanol,practically insoluble in chloroform and ether
Formula Weight 153.61
Percent Purity 99%
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material βine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Betaine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'