missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Betamethasone sodium phosphate

Product Code. 16574504
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-73-5
Molecular Formula C22H28FNa2O8P
Molecular Weight (g/mol) 516.41
InChI Key PLCQGRYPOISRTQ-LWCNAHDDSA-L
IUPAC Name disodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
SMILES [Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O

Specifications

Melting Point 206.0°C to 211.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in chloroform, methanol
Formula Weight 516.41
Chemical Name or Material βmethasone sodium phosphate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'