missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Betamethasone sodium phosphate
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 206.0°C to 211.0°C |
Quantity | 1 g |
Solubility Information | Solubility aq. soln.: slightly soluble. Other solubilities: soluble in chloroform, methanol |
Formula Weight | 516.41 |
Chemical Name or Material | βmethasone sodium phosphate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'