Learn More
Texwipe™ BetaMop™ II
Low contamination properties make this the perfect mop for floor cleaning in critical environments
Brand: Texwipe™ TX716R
315.03 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Refill Mop Head | |
Corrosion | |
For Floor Cleaning in Critical Environments |
Autoclavable | |
BetaMop II | |
Polyester |