missing translation for 'onlineSavingsMsg'
Learn More
Bialaphos sodium, 85%, Thermo Scientific™ Product Code.: 15428309

Bialaphos sodium, 85%, Thermo Scientific™

Product Code. 15428309
100mg
Click to view available options
Quantity:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15428309

Brand: Thermo Scientific Alfa Aesar J66802.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bialaphos sodium (bilanafos), CAS # 35597-43-4, is a tripeptide antibiotic used to confirm gene transfer in plants through use of the bar selection gene.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
• Bialaphos is a bacterial metabolite and an antimicrobial
Application
• Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
• Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71048-99-2
Molecular Formula C11H21N3NaO6P
Molecular Weight (g/mol) 345.268
MDL Number MFCD01674432
InChI Key RTWIRLHWLMNVCC-WQYNNSOESA-M
PubChem CID 118988468
IUPAC Name sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
SMILES CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]

Specifications

Color Orange
Quantity 100mg
Flash Point >110°C (230°F)
UN Number UN2811
Boiling Point 793°C
Solubility Information Soluble in water
Formula Weight 345.26
Percent Purity 85%
Chemical Name or Material Bialaphos sodium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L006 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberJ66802.ME,J66802.MC
Lot NumberP24L007 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog NumberJ66802.ME,J66802.MC
Lot NumberZ20K050 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog NumberJ66802.ME,J66802.MC
Lot NumberY13K059 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ66802.ME,J66802.MC
Lot NumberU16K020 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog NumberJ66802.ME,J66802.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bialaphos sodium, 85%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.