missing translation for 'onlineSavingsMsg'
Learn More

Bicinchoninic acid disodium salt, Thermo Scientific Chemicals

Product Code. 11369778
Click to view available options
Quantity:
2 g
10 g

For determination of Cu and protein assays

For determination of Cu and protein assays.Bicinchoninic acid disodium salt is useful for the determination of copper. It is also used for the determination of protein concentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 979-88-4
Molecular Formula C20H10N2Na2O4
Molecular Weight (g/mol) 388.29
MDL Number MFCD00037500
InChI Key AUPXFICLXPLHBB-UHFFFAOYSA-L
Synonym sodium 2,2'-biquinoline-4,4'-dicarboxylate, 2,2'-biquinoline-4,4-dicarboxylic acid disodium salt, 2,2'-biquinoline-4,4'-dicarboxylic acid, disodium salt, sodium bicinchoninate, 2,2'-biquinoline-4,4'-dicarboxylic acid disodium salt, disodium 2,2'-biquinoline-4,4'-dicarboxylate, bicinchoninic acid disodium salt, bca, 2,2'-biquinoline-4,4'-dicarboxylic acid, sodium salt 1:2
PubChem CID 164763
SMILES [Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1

Specifications

Quantity 10 g
Solubility Information Soluble in water.
IUPAC Name disodium;2-(4-carboxylatoquinolin-2-yl)quinoline-4-carboxylate
Formula Weight 388.29
Physical Form Chunks and/or Powder
Chemical Name or Material Bicinchoninic acid disodium salt
compliance-icons
Product Identifier
  • Bicinchoninic acid, disodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : [2,2'-Biquinoline]-4,4'-dicarboxylic acid, disodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'