missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals BICINE, 99% Product Code.: 11477157

Thermo Scientific Chemicals BICINE, 99%

Product Code. 11477157
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11477157

Brand: Thermo Scientific Chemicals A14957.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicine is widely utilized as a buffering agent for low temperature work. It is used in kinetic study of a quaternary transition-state analogue complex of creatine kinase reaction. It is employed in the thin layer ion exchange chromatography method for protein resolution. It is employed in serum guanase determination by the preparation of stable substrate solution.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N,N-Bis(2-hydroxyethyl)glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Melting Point ∼191°C (decomposition)
pH 4 to 5
Quantity 250 g
Assay Percent Range 99%
Beilstein 1769362
Merck Index 14,1201
Solubility Information Soluble in water.
Formula Weight 163.17
Percent Purity 99%
Chemical Name or Material BICINE

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249643 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog NumberA14957.30,A14957.18,A14957.0B
Lot Number10246309 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14957.30,A14957.18,A14957.0B
Lot Number10241911 Certificate TypeCertificate of Analysis Date20/07/2022 Catalog NumberA14957.30,A14957.18,A14957.0B
Lot Number10206887 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14957.30,A14957.18,A14957.0B
Lot Number10231091 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14957.30,A14957.18,A14957.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals BICINE, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.