missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bicine, 99+%, for biochemistry

Product Code. 10331104
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-Bis(2-hydroxyethyl)glycine, Diethylolglycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Absorbance 0.05 max. (0.5M, water) at 260nm, 0.05 max. (0.5M, water) at 280nm
Melting Point 190.0°C to 192.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Linear Formula (HOCH2CH2)2NCH2CO2H
Packaging Plastic bottle
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, ether, chloroform and, benzene
Formula Weight 163.17
Percent Purity ≥99%
Grade Biochemical
Chemical Name or Material Bicine, For biochemistry
Mostrar más Mostrar menos
Nombre del producto
  • Bicine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'