missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bicine, Fisher BioReagents
£106.00 - £287.00
Chemical Identifiers
CAS | 150-25-4 |
---|---|
Molecular Formula | C6H12NNaO4 |
Molecular Weight (g/mol) | 185.16 |
MDL Number | MFCD00004295 |
InChI Key | MFBDBXAVPLFMNJ-UHFFFAOYSA-M |
Synonym | N, N-bis [2-hydroxyethyl]glycine |
PubChem CID | 8761 |
ChEBI | CHEBI:40957 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]acetic acid |
SMILES | [Na+].OCCN(CCO)CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
10638203
|
Fisher BioReagents
BP2646-100 |
100 g | Poly Bottle | N/A | N/A | N/A | |||||
11453094
|
Fisher BioReagents
BP2646-250 |
250 g | Poly Bottle | N/A | N/A | N/A | |||||
10766965
|
Fisher BioReagents
BP2646-1 |
1 kg | Poly Bottle | N/A | N/A | N/A | |||||
Description
Bicine is one of the zwitterionic amino acids known as Good buffers. Used as a biological buffer and chelating agent.Chemical Identifiers
150-25-4 | |
185.16 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
8761 | |
2-[bis(2-hydroxyethyl)amino]acetic acid |
C6H12NNaO4 | |
MFCD00004295 | |
N, N-bis [2-hydroxyethyl]glycine | |
CHEBI:40957 | |
[Na+].OCCN(CCO)CC([O-])=O |
Specifications
0.100 AU max. at 260nm, 0.100 AU max. at 280nm | |
190°C | |
4.0 to 5.0 | |
C6H12NNaO4 | |
MFCD00004295 | |
N, N-bis [2-hydroxyethyl]glycine | |
[Na+].OCCN(CCO)CC([O-])=O | |
185.16 | |
CHEBI:40957 | |
Bicine |
150-25-4 | |
White | |
Powder or Solid | |
(HOCH2CH2)2NCH2COOH | |
15, 1205 | |
MFBDBXAVPLFMNJ-UHFFFAOYSA-M | |
2-[bis(2-hydroxyethyl)amino]acetic acid | |
8761 | |
163.2 |
Safety and Handling

Recommended Storage : RT
Product Identifier
- Bicine
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bicine, Fisher BioReagents