missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.2.0]hept-2-en-6-one, 97%, Thermo Scientific Chemicals

Product Code. 11460647
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13173-09-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00066201
InChI Key LNLLHUHPGPKRBM-UHFFFAOYSA-N
Synonym bicyclo 3.2.0 hept-2-en-6-one, +/--cis-bicyclo 3.2.0 hept-2-en-6-one, cis-bicyclo 3.2.0 hept-2-en-6-one, 1s,5s-bicyclo 3.2.0 hept-2-en-6-one, +-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-en-6-one, bicyclo 3,2,0 hept-2-ene-6-one, bicyclo 3.2.0 hept-3-en-7-one, z-bicyclo 3.2.0 hepta-2-ene-6-one
PubChem CID 297183
IUPAC Name bicyclo[3.2.0]hept-3-en-7-one
SMILES C1C=CC2C1C(=O)C2

Specifications

Quantity 5 g
UN Number UN1224
Sensitivity Air Sensitive
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material Bicyclo[3.2.0]hept-2-en-6-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.