missing translation for 'onlineSavingsMsg'
Learn More

Bifenazate, MedChemExpress

MedChemExpress Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.

Encompass_Preferred

£60.50 - £168.19

Specifications

Chemical Name or Material Bifenazate
CAS 149877-41-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H20N2O3
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16491616
Encompass_Preferred
View Documents
Medchem Express
HY-119687
500 mg
£113.74
500mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16485676
Encompass_Preferred
View Documents
Medchem Express
HY-119687
100 mg
£60.50
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16431219
Encompass_Preferred
View Documents
Medchem Express
HY-119687
1 g
£168.19
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bifenazate is a carbazate acaricide that control 100% of mites at a concentration of 25 ppm. Bifenazate is a positive allosteric modulator of GABA receptor.
Specifications

Specifications

Bifenazate
Yellow
C17H20N2O3
COC1=CC=C(C2=CC=CC=C2)C=C1NNC(OC(C)C)=O
300.35
Research
Neuroscience-Neuromodulation
149877-41-8
Research
DMSO : ≥ 250 mg/mL (832.36 mM)
300.35
99.65%
Solid
Videos
Safety and Handling

Safety and Handling

H317

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Bifenazate, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.