missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bifonazole, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
Melting Point | 149.0°C |
Loss on Drying | 0.5% max. (1 g, 105°C, 1 h) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 310.39 |
Percent Purity | 98% |
Chemical Name or Material | Bifonazole |
Safety and Handling
Product Identifier
- Bifonazole
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction