missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ BigDye™ Terminator v1.0 & v3.0 5X Sequencing Buffer
This sequencing buffer is optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.
Brand: Applied Biosystems™ 4305603
Additional Details : Weight : 0.01360kg
Description
Optimized for use with BigDye™ Terminator v1.0 or v3.0 cycle sequencing kits.Order Info
10.8mL
Specifications
Sequencing Buffer | |
Store at -15 to -25°C | |
Standard Sequencing | |
Sequencing Buffer | |
Dry Ice |
5 X | |
GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer | |
BigDye™ Terminator, BigDye™ | |
10.8 mL | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.