missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.
£44.69 - £10180.00
Specifications
Buffer | Sequencing Buffer |
---|---|
Concentration | 5X |
Content And Storage | Store at 2°C to 8°C. |
For Use With (Application) | Standard Sequencing |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15768289
|
Applied Biosystems™
4336697 |
1 mL |
£61.75
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15791816
|
Applied Biosystems™
4336699 |
28 mL |
£1248.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15879776
|
Applied Biosystems™
4336701 |
233 mL |
£10180.00
Each |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939182
|
Applied Biosystems™
4336699 |
28 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929182
|
Applied Biosystems™
4336697 |
1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16355292
|
Applied Biosystems™
4336701 |
233 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Optimized for use with BigDye™ Terminator Cycle Sequencing Kits
Order Info
Each
Specifications
Sequencing Buffer | |
Store at 2°C to 8°C. | |
Room Temperature |
5X | |
Standard Sequencing | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.