Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry
Brand: Applied Biosystems™ 4337454
205.74 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Additional Details : Weight : 0.01000kg
Includes: 10 x 20mL Tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 4 tubes M13 (-21) Primer, 4 tubes pGEM Control DNA, 1 x 233mL tube of 5X Sequencing Buffer (may be stored at 4°C)
Description
- Improve the quality of your results for a wide range of sequencing applications
- Optimized for long read lengths
- Better dye mobility characteristics
- Improved performance reading through GT rich regions
- Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
- Enhance your productivity and reduce costs
Order Info
Each
Specifications
200μl tube of BigDye™ Terminator v3.1 Ready Reaction Mix, 1 tube M13 (-21) Primer, 1 tube pGEM Control DNA, and a 1 ml tube of 5X Sequencing Buffer. Store kit at -15°C to -25°C. The BigDye™ sequencing buffer can be stored at 4°C. | |
310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, GeneAmp 9700, Veriti Thermal Cycler, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer | |
BigDye™ Terminator | |
BAC DNA, Lambda DNA, Genomic DNA (Bacterial), Single Stranded DNA, Rolling Circle Amplified Products, Fosmids, PCR Amplicons (50:50 Heterozygotes), Plasmid DNA (≤15Kb), PCR Amplicons |
Kit | |
AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations - 50:50), GC-Rich Sequencing (>65%), Mixed-Base Detection, Long-Read Sequencing, Resequencing, De Novo Sequencing - High Throughput (cDNA), GT-Rich or Difficult Template Sequencing | |
Sequencing Buffer |
For Research Use Only. Not for use in diagnostic procedures.