missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Bile Esculin Agar
Isolate and differentiate enterococci / Group D streptococci and non-Group D streptococci using this selective microbiological medium.
Brand: Thermo Scientific™ PO0169A
16.90 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Straw/ Brown, clear | |
90mm Monoplate | |
Bile Esculin Agar | |
19.5 g ± 1.0 g | |
10/Pk. |
2°C to 10°C | |
United Kingdom | |
Agar | |
Flow Wrapped - Single Layer |