Learn More
Emma Safety Footwear Billy XD Safety Shoes
All purpose safety boot repels acid and oil like an umbrella repels water. Strong, ankle supporting upper fits the foot snugly and comfortably.
Brand: Emma Safety Footwear MM73186844
Additional Details : Weight : 1.95300kg
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Hydro-Tec™ Sanitized Silver for drier, fresher feet
- Sanitized
Specifications
Billy XD | |
Leather | |
44 |