Learn More
Thermo Scientific™ BindIt™ Software, With CD only
Automate customized applications using Thermo Scientific™ BindIt™ Software, a versatile tool for protocol creation and modification.
Brand: Thermo Scientific™ 5189009
Description
Create and store KingFisher purification system protocols using Thermo Scientific™ BindIt™ Software. Once a protocol has been created, the protocol can either be transferred to the Thermo Scientific™ KingFisher™ instrument memory or executed directly from the software. All protocol steps have default parameters which can be changed according to the demands of the application.
Features:- Plates and reagents used are defined in the plate layout
- Due to an increased QC demand, BindIt Software enables the user to generate a status report including the run log, plate layout and step parameters
- Enables the KingFisher Flex to interface with liquid handling, robotics and plate stacking instruments, providing a fully automated solution and the highest possible throughput
Compatible with:
KingFisher, KingFisher mL, KingFisher Flex, KingFisher Duo Prime
Specifications
BindIt Software | |
KingFisher, KingFisher mL, KingFisher Flex, KingFisher Duo Prime | |
One serial or USB port available | |
1 GB RAM |
XVGA monitor with 1024 by 768 resolution | |
CD only | |
Each | |
Windows XP Professional with Service Pack 3, Windows Vista Service with Pack 2, Windows 7 with Service Pack 1, and Windows 8 |