Learn More
Olympus™ Binocular Microscope, Basic Stand, No Illumination, SZ51
Deliver images with superb depth, clarity, and detail. Olympus™ Binocular Microscope, Basic Stand, No Illumination, SZ51 uses dependable, high-performance optics to produce consistent and accurate results and a V-shape optical path to ensure a slim body ideal for equipment integration or for stand alone use.
Brand: Olympus™
Code : NEW
Additional Details : Weight : 6.50000kg
Description
- Binocular head, 45º inclination.
- Interpupillary adjustment 52-76mm.
- Greenough zoom optical system.
- Widefield eyepieces WF10X FN22mm.
- Zoom ratio 5:1.
- 110mm working distance.
- Stand with focusing & ESD capability.
- Black and white stage plate.
Specifications
WF 10X FN22 mm | |
Microscope | |
Binocular | |
0.8X to 4X |