Learn More
Carl Zeiss™ Binocular Microscopes
Brand: Carl Zeiss™ 491206-0001-000
2344.61 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
Primo Vert available as predefined configurations, but additional configurations and accessories are available on request. A number of attractive features are designed to guarantee quick and reliable handling and ease of use:- Automatic light source switch-off (in addition to manual on/off-function): Primo Vert switches off the light automatically after 15min - saving energy and protecting the stand
- Modular illumination: halogen or long lasting LED
- High quality material: the stand is made of metal and all operating elements are coated with a special touch-friendly rubber compound
- Universal phase slider (optional) for all objectives: for the ease of use there is one phase ring (Ph1) available for 10x, 20x and 40x - this means not having to change the position of the phase slider when changing objectives. In short: time-saving and economical
- Increased working distance thanks to easy removal of the condenser, e.g., for roller-bottles
- Objective indicators for fast identification of magnification
- Integrated carrying handle
- Award-winning design The new Primo Vert inverted microscope is focused on the essentials: excellent optics, high-durability quality materials, ease of use and an appealing design. The Primo Vert addresses a wide range of applications: from routine laboratories for live cell inspection to cutting edge research laboratories which require - in addition to research microscopes - compact and reliable routine microscopes for quick and efficient viewing of living cells. Typical applications include cancer and HIV research, human, animal and plant genetics, and cell biology in general.