Learn More
Econix™ Bio-bin™ General/Non Infectious Bins
Use Econix™ Bio-bin™ General/Non Infectious Bins to dispose of single use implements and reduce overfilling issues for your non-sharp clinical waste. Its longer base allows single use lab and surgical instruments to lie flat.
Brand: Econix™ BEXBB30Plain
This item is not returnable.
View return policy
Description
- Autoclavable
- Built in lid and handles
- Temporary and permanent seals
- Waterproof construction
- Space efficient fold flat design
- Stackable post use
Product Content Correction
Product Title
Econix™ Bio-bin™ General/Non Infectious Bins > Capacity: 30L; Color: Plain
Spot an opportunity for improvement?Share a Content Correction