Learn More
Agilent BioTek Synergy LX Multi-Mode Reader

The BioTek Synergy LX multi-mode reader automates many common microplate assays. For Research Use Only. Not for use in diagnostic procedures.
£13758.00 - £18363.00
Specifications
Absorbance Range | 200 to 900 nm |
---|---|
Accuracy | Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Bandwidth | ≤5 nm |
Depth (Metric) | 38.1 cm |
Product Code | Brand | Model | Includes | Height (English) | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Includes | Height (English) | Height (Metric) | Price | Quantity & Availability | ||
17247829
![]() |
Agilent BioTek
SLXA-SN |
SLXA | Absorbance | 12 in. | 30.5 cm |
£13758.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17267829
![]() |
Agilent BioTek
SLXATS-SN |
SLXATS | Absorbance; Touchscreen | 15 in. | 38.1 cm |
£15294.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17287829
![]() |
Agilent BioTek
SLXF-SN |
SLXF | Fluorescence; Luminescence | 12 in. | 30.5 cm |
£13758.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17247839
![]() |
Agilent BioTek
SLXFTS-SN |
SLXFTS | Fluorescence; Luminescence; Touchscreen | 15 in. | 38.1 cm |
£15294.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17207839
![]() |
Agilent BioTek
SLXFA-SN |
SLXFA | Absorbance; Fluorescence; Luminescence | 12 in. | 30.5 cm |
£16826.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17227839
![]() |
Agilent BioTek
SLXFATS-SN |
SLXFATS | Absorbance; Fluorescence; Luminescence; Touchscreen | 15 in. | 38.1 cm |
£18363.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability
The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.
Warranty
12 Months
Specifications
200 to 900 nm | |
CE and TUV marked. RoHS compliant. | |
38.1 cm | |
24 sec. (96 wells), 76 sec. (384 wells) | |
Monochromator Bandpass | |
50/60 Hz | |
Multi-mode Microplate Reader | |
0.001 OD | |
2 | |
1 year | |
27 lb. | |
15 in. | |
Photodiode, PMT | |
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning |
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD | |
≤5 nm | |
15 in. | |
Linear, Orbital, Double Orbital | |
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability | |
<1% from 0 to 2.5 OD | |
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) | |
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP | |
English | |
100/240 V | |
12.3 kg | |
38.1 cm | |
Xenon Flash |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.