missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (30%, 39:1), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar J60412.K2

143.45 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : NEW

Additional Details : Weight : 1.00000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III

 View more versions of this product

Product Code. 15433189

  • £151.00 / 1 L
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acrylamide/Bisacrylamide 39:1
30% soln
1 L
Liquid
Colorless
UN3426
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only