Learn More
BioAcryl-P (40%, 29:1), Thermo Scientific Chemicals
BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.
Brand: Thermo Scientific Alfa Aesar J63079.AP
83.41 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00050kg
Description
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acrylamide/Bisacrylamide 29:1 | |
40% soln | |
500 mL | |
UN3426 |
Liquid | |
Colorless | |
MFCD00080848 | |
It is soluble in water. |
RUO – Research Use Only