Learn More
BioAcryl-P (40%, 29:1)
Description
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
- Acrylamide is a crystalline amide that forms polymer structures at room temperature
- Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation
Application
- Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
Specifications
Specifications
| Chemical Name or Material | Acrylamide/Bisacrylamide 29:1 |
| Physical Form | Liquid |
| Concentration | 40% soln |
| Color | Colorless |
| Quantity | 1 L |
| MDL Number | MFCD00080848 |
| UN Number | UN3426 |
| Solubility Information | It is soluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.