missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Biobased Flat Tray

Biobased Flat Tray has dimensions suitable for refrigerators.

Brand:  Gilac™ G152421

 View more versions of this product

Product Code. 17948871

  • £17.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

  • Flat tray in biobased material.
  • Dimensions suitable for refrigerators.
  • Flat tray can be used as:
    • defrosting trays thanks to its dividing grid that separates the product from the defrosting water.
    • preparation container for products that drip.
    • ingredients containers.
    • containers for preserving comparison samples.
    • Fermenting and decanting containers.
  • Nesting and stacking with lid. Traceability management thanks to a contents identification label (IML).
  • 3 volumes available: 3 L, 5 L, 8 L.
  • Biobased flat trays are white and dye-free.
  • Biobased material is made from natural renewable resources instead of fossil resources.
  • As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety.
Specifications

Specifications

8 L
75 mm
HDPE
335 mm
3 in.
2.1 gal.
White
485 mm
Laboratory Tray
19.1 in.
13.2 in.
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.