Learn More
Gilac™ Biobased Rectangular Dividing Dough Container
Biobased Rectangular Dividing Dough Container is highly resistant to changes in temperature, highly flexible and shock-resistant.
Brand:  Gilac™ G155491
Additional Details : Weight : 1.15500kg
Product Code. 17918821
  • £20.26 / Each
In Stock
to see stock.
Description
Description
  • Rectangular dividing dough container in biobased material.
  • Highly resistant to changes in temperature.
  • Highly flexible and shock-resistant.
  • Compatible with racking trolleys.
  • Rectangular biobased dividing dough container stackable (with lid) and nestable.
  • Rectangular biobased dividing dough container is white and dye-free.
  • Biobased material is made from natural renewable resources instead of fossil resources.
  • As a responsible French manufacturer, GILAC is now offering alternatives to standard materials, whilst maintaining an optimal level of quality and food safety.
Specifications
Specifications
Storage Bin
White
5.3 gal.
HDPE
Rectangular
20 L