Learn More
Thermo Scientific™ BioBasic™ 18 HPLC Columns
Use this 300Å wide-pore silica for the separation of small to medium proteins and other biomolecules.
£363.32 - £4182.56
Specifications
Particle Size | 5 μm |
---|---|
pH | 2 to 9 |
Pore Size | 300 Å |
Surface Area | 100 m²/g |
Temperature | 60°C |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10434773
|
Thermo Scientific™
72105-254630 |
5 μm | 4.6 mm | 250 mm |
£727.62
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10151213
|
Thermo Scientific™
72105-252130 |
5 μm | 2.1 mm | 250 mm |
£806.78
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
16318616
|
Thermo Scientific™
72105-259270A |
5 μm | 20 mm | 250 mm |
£4182.56
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
10097534
|
Thermo Scientific™
72105-151030 |
5 μm | 1 mm | 150 mm |
£793.87
Each |
In Stock
Log in to see stock. |
|||
10435553
|
Thermo Scientific™
72105-051030 |
5 μm | 1 mm | 50 mm |
£769.50
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10181113
|
Thermo Scientific™
72105-152130 |
5 μm | 2.1 mm | 150 mm |
£724.62
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10293113
|
Thermo Scientific™
72105-102130 |
5 μm | 2.1 mm | 100 mm |
£688.48
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
10475173
|
Thermo Scientific™
72105-052130 |
5 μm | 2.1 mm | 50 mm |
£630.27
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
10502645
|
Thermo Scientific™
72105-154630 |
5 μm | 4.6 mm | 150 mm |
£690.49
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10160703
|
Thermo Scientific™
72105-101030 |
5 μm | 1 mm | 100 mm |
£835.41
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
11558310
|
Thermo Scientific™
72105-014001 |
5 μm | 4 mm | 10 mm |
£363.32
Pack of 4 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
11548310
|
Thermo Scientific™
72105-012101 |
5 μm | 2.1 mm | 10 mm |
£402.86
Pack of 4 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10448674
|
Thermo Scientific™
72105-011001 |
5 μm | 1 mm | 10 mm |
£403.76
Pack of 4 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
10735706
|
Thermo Scientific™
72105-251030 |
5 μm | 1 mm | 250 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.
- 300Å pore size for maximum performance with biomolecules
- High peak capacity stationary phase
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Specifications
5 μm | |
300 Å | |
60°C | |
C18 | |
Reversed Phase | |
Reversed Phase |
2 to 9 | |
100 m²/g | |
L1 | |
9% | |
Yes |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.