Learn More
Thermo Scientific™ BioBasic™ AX HPLC Columns
Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.
Brand: Thermo Scientific™ 73105-051030
Description
Use Thermo Scientific™ BioBasic™ AX LC Columns for the separation of proteins, peptides, other anionic species and highly polar molecules.
- 5μm particle size, 300Å pore size
- Proprietary, protein-friendly, ion exchange ligands of polyethyleneimine (PEI) bound to pure deactivated silica
- Low hydrophobicity
- Capacity of 0.22mEq/g; 3% carbon load
- Use for normal phase and hydrophilic interaction chromatography (HILIC)
- Stable under demanding pH conditions (2 to 8 pH range)
Specifications
Analytical Column | |
Low | |
5800 psi (400 bar) | |
2 to 8 | |
100 m²/g | |
N/A | |
BioBasic | |
3% | |
No | |
Bio-LC |
1 mm | |
50 mm | |
5 μm | |
300 Å | |
60°C | |
Polyethyleneimine | |
Spherical, Fully Porous, Ultrapure Silica | |
Mixed Mode | |
Normal Phase, HILIC, Ion Exchange |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.