Learn More
Chemo Protective Sterile Sleevecovers
Manufactured for use in ISO Class 4 environments, BioClean-C™ Sterile Protective Sleeve Covers are chemotherapy protective sleeves, the seams of which are covered with protective tape sterilized with gamma irradiation.
Brand:  Nitritex S-BCSC
Product Code. 15268278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Ultrasonically bonded seams covered with protective tape
  • ISO 9001:2008 certification
  • Sterilized with gamma irradiation