Learn More
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots Product Code.: 17841009
Add to basket
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots
Product Code. 17841009 Shop All Ansell Products
Sterility: Sterile
Click to view available options
Sterility: Sterile
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17841009
Brand: Ansell SBDFCXS
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Cleanroom Coverall made of CleanTough™ fabric for lightweight, low-linting body protection with front zip and protective flap to provide contaminant and liquid splash barrier.
  • Reduced contamination risks:  Lightweight CleanTough™ material means the BioClean-D Coverall with Hood and Integrated Boots – Sterile S-BDFC is low-linting and ISO cleanroom-compliant, reducing contamination risks
  • Optimized protection: This sterile coverall features a front zip with a protective flap, resisting liquid splashes and chemicals*, while thumb loops and an elasticated hood, back, cuffs and ankles ensure a secure fit
  • Enhanced features: This antistatic coverall features integrated boots and slip-resistant soles, for added workplace safety in laboratory settings
  • Sterility assurance level: Special packaging eliminates cross contamination, while gamma irradiation sterilization ensures a minimum SAL of 10-6
  • Contact customer service for chemotherapy drug permeation times and recommendations.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Flashspun Polyethylene
Color White
Disposable Single-use
Waist Style Standard
Ankle Style Attached Boots, Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Sterile
Resistance Anti-static, Chemicals, Liquid, Particle
Collar Type Hooded
Certifications/Compliance (Europe) ASTM F739, CE 0598, EN 1149-5:2008, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 13688:2013, ISO 11137-1:2006, EN ISO 14325, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009
Latex Free No
No. of Pockets None
For Use With (Application) Blending, compounding solids or liquids, Crime Scene Investigation (CSI), Forensics, Handling of Powders, Aseptic Filling and Labelling, Cleanroom Cleaning and Preparing, Medical Device Manufacturing, Biotechnologies Blending Operations, Handling Active Material, Maintaining Equipment and Instruments
Cleanroom Class Class 10 (ISO 4)
Product Title
Ansell BioClean-D™ Sterile cleanroom Coverall with Hood and integrated Boots >