missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens

Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.

£4.18 - £655.00

Specifications

Length (English) 5.5 in.
Length (Metric) 140 mm
View More Specs

Products 6
Product Code Brand Color Description Price Quantity & Availability  
Product Code Brand Color Description Price Quantity & Availability  
15298308
View Documents
Nitritex™
S-BPTB-BK1
Black Bagged This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15994841
View Documents Promotion Details
Nitritex™
S-BPFP-BK1
Black Double bagged
£31.95
Pack of 10
In Stock
to see stock.
Edge
Add to basket
 
15365905
View Documents Promotion Details
Nitritex™
S-BPFP-BL1
Blue Double bagged, Pack of 10
£31.95
Pack of 10
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
15278308
View Documents
Nitritex™
S-BPFP-BL1
Blue Double bagged This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15218318
View Documents
Nitritex™
S-BPTB-RD1
Red Bagged This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15904851
View Documents
Nitritex™
S-BPFP-RD1_clean
Red Double bagged This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Super permanent ink in black, blue, or red
  • Fade and water resistant
  • Quick drying
  • Non-toxic
  • Alcohol resistant labeling
  • Autoseal ink prevents drying
  • Store in a dry, cool place away from direct sunlight and fluorescent lighting
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Processed in an NEBB certified ISO Class 4 cleanroom environment
Specifications

Specifications

5.5 in.
140 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.