missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ BioClean™ Sterile Nitrile RABS/Isolator Sleeve
BIOPHARMA

Product Code. 30002923
Click to view available options
Port Size (English):
12 to 14 in.
6 to 8 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

The BioClean™ Sterile Nitrile RABS/Isolator Sleeve is manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.

  • 100% water leak tested (before being guillotined).
  • Suitable for use with a wide range of cuff systems.
  • Triple bagged.
  • Autoclave compatible - sleeve only.
TRUSTED_SUSTAINABILITY

Specifications

ASTM F739 No
Certifications/Compliance (Europe) ISO 11137-1:2006, EN 420:2003 + A1:2009
Chlorinated Yes
Cleaning Procedure DI Washing
Cleanroom Class Class 10 (ISO 4)
Content And Storage Store in a dry, cool place (<40°C) away from direct sunlight and fluorescent light.
Description One piece per sealed inner PE bag, one inner bag per sealed second inner PE bag, one second inner bag per sealed outer PE bag, 20 outer bags per lined inner white Correx polyethylene box
Accelerator Free No
Applications Laboratory
Color White
Latex Free Yes
Material Nitrile, Latex Dipped
Style Beaded Cuff
Thickness (English) 20 mil
Thickness (Metric) 0.50 mm
For Use With (Application) Pharmaceutical Manufacturing, Laboratory Research and Development, Raw Material and Preparation, Biotechnology Manufacturing, Sterile Processing Department, Semiconductor Manufacturing
Gender Unisex
Packaging Type PE Bags, Correx Polyethylene Box
Port Size (English) 6 to 8 in.
Quantity 20 Pk.
Series GSL
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'