missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ 96-Well, Gelatin-Treated, Flat-Bottom Microplate
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Packaging:
5/Cs
50/Cs
Description
Corning BioCoat Gelatin cultureware provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of vascular endothelial cells, muscle, embryonic stem (ES) cells, and F9 teratocarcinoma cells. It is also suitable for promoting adhesion of transfected cell types. Gelatin is a heterogeneous mixture of water-soluble proteins derived through the hydrolysis of Collagen.
Applications include:
- Promotion of cell attachment and spreading
- Culture of normal and transfected F9 teratocarcinoma cells for gene expression studies
- Culture of HUVEC for E-Selectin7 expression and VEGF induction8
Source
Quality Control
- Tested for ability to promote proliferation of HUVECs
- Tested and found negative for bacteria and fungi
Storage
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Format | Standard |
Lid | With Lid |
Content And Storage | 4°C to 30°C |
Culture Area | 0.32 cm2 |
Array | 8 x 12 |
Packaging | 50/Cs |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Sterility | Sterile |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'